Learn More
Cayman Chemical N-Nonyldeoxynojirimycin, Purity: ≥98%, CAS: 81117-35-3, 10 mg
SDP
Catalog No. 502988829
Encompass_Preferred
An inhibitor of α-glucosidase and α-1,6-glucosidase (IC50s = 0.42 and 8.4 μM, respectively); increases hepatic glycogen levels in fasted mice and gastrocnemius muscle glycogen levels in both fasted and fed mice at 250 mg/kg per day; increases β-glucosidase activity by 1.65-fold in patient-derived fibroblasts homozygous for the N370S Gaucher disease mutation when added to the culture medium at 5 μM, but inhibits β-glucosidase activity at >60 μM; DNJ inhibits viral replication in BHK-21 cells JEV or DEN-2); has protective effects in a mouse model of lethal JEV challenge at 200 mg/kg per day
Catalog No. 50-298-8829 Supplier Cayman Chemical Supplier No. 2969810 MG
An inhibitor of α-glucosidase and α-1,6-glucosidase (IC50s = 0.42 and 8.4 μM, respectively); increases hepatic glycogen levels in fasted mice and gastrocnemius muscle glycogen levels in both fasted and fed mice at 250 mg/kg per day; increases β-glucosidase activity by 1.65-fold in patient-derived fibroblasts homozygous for the N370S Gaucher disease mutation when added to the culture medium at 5 μM, but inhibits β-glucosidase activity at >60 μM; DNJ inhibits viral replication in BHK-21 cells JEV or DEN-2); has protective effects in a mouse model of lethal JEV challenge at 200 mg/kg per day