Learn More
Medchemexpress LLC N-Nornuciferine | 4846-19-9 | 1 ML
SDP
Catalog No. 5000421674
Encompass_Preferred
N-Nornuciferine is an aporphine alkaloid derived from lotus leaf. It is known for its significant inhibitory activity against the CYP2D6 enzyme, with IC50 and Ki values of 3.76 μM and 2.34 μM, respectively.
  • Derived from lotus leaf
  • Selectively inhibits CYP2D6 activity
  • Shows weak or no inhibition of other P450 isoenzymes (CYP2C19, CYP3A4, CYP2E1, CYP2C9)
  • Competitively inhibits dextromethorphan o-demethylation
Catalog No. 50-004-21674 Supplier Medchemexpress LLC Supplier No. HYN212910MM/1ML
N-Nornuciferine is an aporphine alkaloid derived from lotus leaf. It is known for its significant inhibitory activity against the CYP2D6 enzyme, with IC50 and Ki values of 3.76 μM and 2.34 μM, respectively.
  • Derived from lotus leaf
  • Selectively inhibits CYP2D6 activity
  • Shows weak or no inhibition of other P450 isoenzymes (CYP2C19, CYP3A4, CYP2E1, CYP2C9)
  • Competitively inhibits dextromethorphan o-demethylation