Learn More
Medchemexpress LLC HY-N2129 5mg Medchemexpress, N-Nornuciferine CAS:4846-19-9 Purity:>98%
SDP
Catalog No. 501933887
Encompass_Preferred
Medchemexpress, HY-N2129 5mg N-Nornuciferine CAS:4846-19-9 N-Nornuciferine is an aporphine alkaloid in lotus leaf that significantly inhibits CYP2D6 with IC 50 and K i of 3.76 and 2.34 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3887 Supplier Medchemexpress LLC Supplier No. HYN21295MG
Medchemexpress, HY-N2129 5mg N-Nornuciferine CAS:4846-19-9 N-Nornuciferine is an aporphine alkaloid in lotus leaf that significantly inhibits CYP2D6 with IC 50 and K i of 3.76 and 2.34 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.