Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,O-bis[(1,1-dimethylethoxy)carbonyl]-L-tyrosine | 20866-48-2 | MFCD00038856 | 98.0% | 381.42 g/mol | C19H27NO7 | 25 G
SDP

Catalog No. 5000208845
Encompass_Preferred

Boc-protected tyrosine derivative used as a building block in peptide and small-molecule synthesis. Supplied as a powder for research use only.

  • Used as an N- and O-protected amino acid for peptide synthesis.
  • Chemical formula C19H27NO7 and molecular weight 381.42 g/mol.
  • CAS number 20866-48-2 for unambiguous identification.
  • Available in multiple package sizes, including 25 G.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-08845 Supplier Medchemexpress LLC Supplier No. 25GHYW048332
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-protected tyrosine derivative used as a building block in peptide and small-molecule synthesis. Supplied as a powder for research use only.

  • Used as an N- and O-protected amino acid for peptide synthesis.
  • Chemical formula C19H27NO7 and molecular weight 381.42 g/mol.
  • CAS number 20866-48-2 for unambiguous identification.
  • Available in multiple package sizes, including 25 G.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.