Learn More
Matrix Scientific N,O-Bis(trimethylsilyl)acetamide, 10416-59-8, MFCD00008270, 100g
Supplier: Matrix Scientific 074838100G
Molecular Formula C8H21NOSi2, Purity 95+%, Molecular Weight 203.43, Melting Point 24°, Boiling Point 71-73/35mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.