Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,O-Bis(Trimethylsilyl)Acetamide

Catalog No. 12891025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-891-025ML 25 mL
12-891-010ML 10 mL
12-891-01L 1 L
12-891-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 12-891-025ML Supplier Sigma Aldrich Supplier No. 12891025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the regioselective desulfation of polysaccharide sulfates. Also used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

Specifications

CAS 10416-59-8
Density 0.832 g/mL at 20°C
Boiling Point 71°C to 73°C35 mmHg, lit.)
Molecular Formula C8H21NOSi2
Refractive Index n20/D 1.417
Linear Formula CH3C[=NSi(CH3)3]OSi(CH3)3
MDL Number MFCD00008270
Quantity 25 mL
Synonym BSA
Molecular Weight (g/mol) 203.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.