Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)acetamide, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020278 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0278 5 mL
11-101-8849 10 x 1 mL
11-101-8607 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-0278 Supplier MilliporeSigma Supelco Supplier No. 152695ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,O-Bis(trimethylsilyl)acetamide is an excellent silylation reagent. For some sterically hindered primary or secondary nitro compounds, N,O-bis(trimethylsilyl)acetamide and BSTFA achieve better results than normal silylation conditions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00008270
Synonym BSA

Specifications

Boiling Point 71°C to 73°C (35 mmHg)
CAS 10416-59-8
Quantity 5 mL
Linear Formula CH3 C[=NSi(CH3)3 ]OSi(CH3)3
Flash Point 44°C
Formula Weight 203.43
Percent Purity ≥98.5% (GC)
Grade GC Derivatization
Density 0.832 g/mL (at 20°C)
RTECSNumber AK3000000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.