Learn More
Medchemexpress LLC N,O-bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane | 25561-30-2 | MFCD00008269 | 98.0% | 257.40 g/mol | C8H18F3NOSi2 | 25G
SDP
Catalog No. 5000235444
Encompass_Preferred
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent used to convert polar functional groups into trimethylsilyl derivatives, improving volatility and thermal stability for gas chromatography and GC-MS analysis. Supplied for research use, it is typically a colorless to light yellow liquid and is provided with documentation to support analytical workflows.
  • Derivatizes hydroxyl, carboxyl, and amino functional groups.
  • Improves volatility and thermal stability for GC and GC-MS analysis.
  • Compatible with common chromatographic workflows and instruments.
  • High purity (98.0%) for reproducible analytical results.
  • Available in multiple packaging sizes to suit routine or bulk use.
  • Documentation available for quality control, including SDS, COA, and NMR.
Catalog No. 50-002-35444 Supplier Medchemexpress LLC Supplier No. HY12379825G
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent used to convert polar functional groups into trimethylsilyl derivatives, improving volatility and thermal stability for gas chromatography and GC-MS analysis. Supplied for research use, it is typically a colorless to light yellow liquid and is provided with documentation to support analytical workflows.
  • Derivatizes hydroxyl, carboxyl, and amino functional groups.
  • Improves volatility and thermal stability for GC and GC-MS analysis.
  • Compatible with common chromatographic workflows and instruments.
  • High purity (98.0%) for reproducible analytical results.
  • Available in multiple packaging sizes to suit routine or bulk use.
  • Documentation available for quality control, including SDS, COA, and NMR.