Learn More
American Radiolabeled Chemicals Inc N-OCTA-D-ER-DSPH-3H 50UC
SDP
Catalog No. 5090104560
Encompass_Preferred
N-Octanoyl-D-erythro-dihydrosphingosine-4,5-3H 50uCi in ethanol, 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 413.5, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-04560 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 060150UC
N-Octanoyl-D-erythro-dihydrosphingosine-4,5-3H 50uCi in ethanol, 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 413.5, Shipped in Dry Ice, Exclusive