Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC n-octacosane-d58 | 16416-33-4 | MFCD00190493 | 99.1% | 453.12 g/mol | C28D58 | 50 MG
SDP

Catalog No. 5000282190
Encompass_Preferred

Deuterium-labeled octacosane (n-octacosane-d58) is an isotope-labeled straight-chain hydrocarbon supplied as a white to off-white solid. It serves as an internal standard, tracer, or reference material for gas chromatography-mass spectrometry and other analytical or metabolic studies requiring a heavy-isotope analogue. The material offers high isotopic and chemical purity and specific storage recommendations for long-term stability.

  • Isotope-labeled internal standard for GC/MS and LC/MS applications.
  • Molecular formula C28D58 and molecular weight 453.12 g/mol.
  • Purity 99.1% for reliable quantitation.
  • Solid, white to off-white form for easy handling.
  • Recommended storage: powder at -20°C for long term, 4°C for shorter term.

Catalog No. 50-002-82190 Supplier Medchemexpress LLC Supplier No. HYW272217S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled octacosane (n-octacosane-d58) is an isotope-labeled straight-chain hydrocarbon supplied as a white to off-white solid. It serves as an internal standard, tracer, or reference material for gas chromatography-mass spectrometry and other analytical or metabolic studies requiring a heavy-isotope analogue. The material offers high isotopic and chemical purity and specific storage recommendations for long-term stability.

  • Isotope-labeled internal standard for GC/MS and LC/MS applications.
  • Molecular formula C28D58 and molecular weight 453.12 g/mol.
  • Purity 99.1% for reliable quantitation.
  • Solid, white to off-white form for easy handling.
  • Recommended storage: powder at -20°C for long term, 4°C for shorter term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.