Learn More
AdipoGen N-Octadecanoyl-Nile Blue
SDP
Supplier:  AdipoGen CDXO0027M100
Encompass_Preferred
Chemical. CAS 125829-24-5. Formula C38H53N3O2. MW 583.85. Synthetic. This membrane probe has been used as an optode sensor for sodium and other ions. It was used to set up optical dihydrogen phosphate-selective sensors better suited for operation at physiological pH than their ion selective electrode counterparts.
Catalog No. 50-256-0343