Learn More
AdipoGen N-Octadecanoyl-Nile Blue, 125829-24-5, MFCD00213935, 10 mg
SDP
Supplier:  AdipoGen CDXO0027M010
Encompass_Preferred
MF: C38H53N3O2, MW: 583.85, ≥98%, Appearance: Violet powder. This membrane probe has been used as an optode sensor for sodium and other ions. It was used to set up optical dihydrogen phosphate-selective sensors better suited for operation at physiological pH than their ion selective electrode counterparts.
Catalog No. 50-117-7656