Learn More
Strem, An Ascensus Company CAS# 4724-47-4. 1g. n-Octadecylphosphonic acid, min. 97% ODPA. MFCD00015838
SDP
Catalog No. 5090110273
Encompass
CAS# 4724-47-4. 1g. n-Octadecylphosphonic acid, min. 97% ODPA. MFCD00015838. Molecular Weight: 334.47. Molecular Formula: CH3(CH2)17P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-3510. http://www.strem.com/catalog/v/15-3510/
Catalog No. 50-901-10273 Supplier Strem, An Ascensus Company Supplier No. 1535101G
CAS# 4724-47-4. 1g. n-Octadecylphosphonic acid, min. 97% ODPA. MFCD00015838. Molecular Weight: 334.47. Molecular Formula: CH3(CH2)17P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-3510. http://www.strem.com/catalog/v/15-3510/