Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(octahydrobenzo[e]pyrido[1,2-a]diazepin-8-yl)thiophene-2-sulfonamide | 1193340-31-6 | 98.0% | 10 MG
SDP

Catalog No. 5000230745
Encompass_Preferred

AN-12-H5 is a small-molecule antienterovirus research compound that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). It is supplied as a solid or as a DMSO solution and is intended for laboratory research use only.

  • Targets viral replication of PV and EV71.
  • High purity (≈98%).
  • Soluble in DMSO (100 mg/mL); ultrasonic recommended.
  • Provided as solid or pre-dissolved DMSO solution for convenient use in assays.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-30745 Supplier Medchemexpress LLC Supplier No. HY12090010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AN-12-H5 is a small-molecule antienterovirus research compound that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). It is supplied as a solid or as a DMSO solution and is intended for laboratory research use only.

  • Targets viral replication of PV and EV71.
  • High purity (≈98%).
  • Soluble in DMSO (100 mg/mL); ultrasonic recommended.
  • Provided as solid or pre-dissolved DMSO solution for convenient use in assays.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.