Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octane-d{18}, 99% (Isotopic)

Catalog No. AA4233906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4233906 5 g
AA4233903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4233906 Supplier Thermo Scientific Chemicals Supplier No. 04233906
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17252-77-6 | C8H18 | 132.342 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17252-77-6
Molecular Formula C8H18
Molecular Weight (g/mol) 132.342
MDL Number MFCD00037626
InChI Key TVMXDCGIABBOFY-VAZJTQEUSA-N
Synonym octane-d18, 2h18 octane, n-octane-d18, 2 h?? octane, octane-d18, 98 atom % d, n-octane-d isotopic 1g
PubChem CID 519375
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecadeuteriooctane
SMILES CCCCCCCC

Specifications

Physical Form Liquid
Quantity 5 g
Density 0.815 g/mL
Flash Point 15.6°C (60°F)
Melting Point -57°C
Boiling Point 126°C
UN Number UN1262
Formula Weight 132.34
Percent Purity 99%
Chemical Name or Material n-Octane-d{18}, Isotopic
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H224-H304-H315-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber 241-285-3
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.