Learn More
Medchemexpress LLC N-Octanoyl-D-erythro-sphingosine | 74713-59-0 | 98.0% | 425.69 | 10 G
SDP
Catalog No. 5000345438
Encompass_Preferred
C8-Ceramide (N-Octanoyl-D-erythro-sphingosine) is a cell-permeable analog of naturally occurring ceramides. It possesses anti-proliferation properties and acts as a potent chemotherapeutic agent. C8-Ceramide stimulates dendritic cells to promote T cell responses upon virus infections and induces slight activation of protein kinase (PKC) in vitro.
  • Cell-permeable analog of naturally occurring ceramides
  • Exhibits anti-proliferation properties
  • Acts as a potent chemotherapeutic agent
  • Stimulates dendritic cells to promote T cell responses upon virus infections
  • Induces slight activation of protein kinase (PKC) in vitro
Catalog No. 50-003-45438 Supplier Medchemexpress LLC Supplier No. HY10839110MM/1ML
C8-Ceramide (N-Octanoyl-D-erythro-sphingosine) is a cell-permeable analog of naturally occurring ceramides. It possesses anti-proliferation properties and acts as a potent chemotherapeutic agent. C8-Ceramide stimulates dendritic cells to promote T cell responses upon virus infections and induces slight activation of protein kinase (PKC) in vitro.
  • Cell-permeable analog of naturally occurring ceramides
  • Exhibits anti-proliferation properties
  • Acts as a potent chemotherapeutic agent
  • Stimulates dendritic cells to promote T cell responses upon virus infections
  • Induces slight activation of protein kinase (PKC) in vitro