Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-octanoyl-DL-homoserine lactone | 106983-30-6 | MFCD01862915 | 99.2% | 227.30 | C12H21NO3 | 25MG
SDP

Catalog No. 5000238519
Encompass_Preferred

N-Octanoyl-DL-homoserine lactone is an N-acyl homoserine lactone used as a bacterial quorum-sensing signal in microbiology and chemical biology research. It is supplied as a solid suitable for assay work, receptor studies, and signaling experiments.

  • Solid white to off-white powder.
  • Molecular formula C12H21NO3; molecular weight 227.30.
  • High purity (99.2%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Stable when stored sealed: powder -80°C (2 years) or -20°C (1 year).
  • Offered in small research pack sizes appropriate for laboratory use.

Catalog No. 50-002-38519 Supplier Medchemexpress LLC Supplier No. HY12423725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Octanoyl-DL-homoserine lactone is an N-acyl homoserine lactone used as a bacterial quorum-sensing signal in microbiology and chemical biology research. It is supplied as a solid suitable for assay work, receptor studies, and signaling experiments.

  • Solid white to off-white powder.
  • Molecular formula C12H21NO3; molecular weight 227.30.
  • High purity (99.2%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Stable when stored sealed: powder -80°C (2 years) or -20°C (1 year).
  • Offered in small research pack sizes appropriate for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.