Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-octanoyl-DL-homoserine lactone | 106983-30-6 | MFCD01862915 | 99.2% | 227.30 g·mol⁻¹ | C12H21NO3 | 10MG
SDP

Catalog No. 5000238467
Encompass_Preferred

N-Octanoyl-DL-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum-sensing signal molecule used in microbiology and chemical biology to investigate bacterial cell-to-cell communication. It is supplied as a white to off-white solid reagent for research applications.

  • Used to study quorum-sensing and AHL-mediated signaling pathways.
  • White to off-white solid form.
  • Molecular formula C12H21NO3; molecular weight 227.30 g·mol⁻¹.
  • Purity 99.2% (HPLC).
  • Available in small research pack sizes such as 5 mg, 10 mg, and 25 mg.
  • Intended for laboratory research use.

Catalog No. 50-002-38467 Supplier Medchemexpress LLC Supplier No. HY12423710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Octanoyl-DL-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum-sensing signal molecule used in microbiology and chemical biology to investigate bacterial cell-to-cell communication. It is supplied as a white to off-white solid reagent for research applications.

  • Used to study quorum-sensing and AHL-mediated signaling pathways.
  • White to off-white solid form.
  • Molecular formula C12H21NO3; molecular weight 227.30 g·mol⁻¹.
  • Purity 99.2% (HPLC).
  • Available in small research pack sizes such as 5 mg, 10 mg, and 25 mg.
  • Intended for laboratory research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.