Learn More
Medchemexpress LLC N-octanoyl-DL-homoserine lactone | 106983-30-6 | MFCD01862915 | 99.2% | 227.30 g·mol⁻¹ | C12H21NO3 | 5MG
SDP
Catalog No. 5000238631
Encompass_Preferred
N-Octanoyl-DL-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum-sensing signal used in microbiology and biochemical research to study bacterial cell-to-cell communication and AHL-mediated gene regulation. The compound is supplied as a white to off-white solid (typical package 5 MG), with formula C12H21NO3, molecular weight 227.30 g·mol⁻¹, and high reported purity.
Catalog No. 50-002-38631 Supplier Medchemexpress LLC Supplier No. HY1242375MG
Add to Cart
Add to Cart
N-Octanoyl-DL-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum-sensing signal used in microbiology and biochemical research to study bacterial cell-to-cell communication and AHL-mediated gene regulation. The compound is supplied as a white to off-white solid (typical package 5 MG), with formula C12H21NO3, molecular weight 227.30 g·mol⁻¹, and high reported purity.