Learn More
TARGETMOL CHEMICALS INC N-octanoyl-L-Homoserine lacton
SDP
Catalog No. 502880083
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-octanoyl-L-Homoserine lactone (C8-HSL) is a diffusible exogenous Yersinia pestis population-sensing molecule that modulates LcrV virulence factors, controls gene expression by participating in population sensing, and affects cellular metabolism.N-octanoyl-L-Homoserine lactone is used to study cystic fiber Infection. Purity 99.64%
Catalog No. 50-288-0083 Supplier TARGETMOL CHEMICALS INC Supplier No. T3774425MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-octanoyl-L-Homoserine lactone (C8-HSL) is a diffusible exogenous Yersinia pestis population-sensing molecule that modulates LcrV virulence factors, controls gene expression by participating in population sensing, and affects cellular metabolism.N-octanoyl-L-Homoserine lactone is used to study cystic fiber Infection. Purity 99.64%