Learn More
Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | 99.4% | 227.30 | C12H21NO3 | 50MG
SDP
Catalog No. 5000236892
Encompass_Preferred
N-octanoyl-L-homoserine lactone is a bacterial quorum-sensing signaling molecule (an N-acyl homoserine lactone, C8-HSL) used as an analytical standard and research reagent for studies of bacterial communication, quorum sensing pathways, and analytical method development. It is provided at high purity for use in biochemical and microbiology assays.
  • High purity suitable for analytical and biological assays.
  • Molecular formula and weight provided for experimental reference.
  • Available in small-scale quantities and ready-to-use solution formats.
  • Compatible with HPLC, LC-MS, and microbiological assays.
  • Can be reconstituted in common solvents such as DMSO for immediate use.
Catalog No. 50-002-36892 Supplier Medchemexpress LLC Supplier No. HY124237A50MG
N-octanoyl-L-homoserine lactone is a bacterial quorum-sensing signaling molecule (an N-acyl homoserine lactone, C8-HSL) used as an analytical standard and research reagent for studies of bacterial communication, quorum sensing pathways, and analytical method development. It is provided at high purity for use in biochemical and microbiology assays.
  • High purity suitable for analytical and biological assays.
  • Molecular formula and weight provided for experimental reference.
  • Available in small-scale quantities and ready-to-use solution formats.
  • Compatible with HPLC, LC-MS, and microbiological assays.
  • Can be reconstituted in common solvents such as DMSO for immediate use.