Learn More
Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | 99.4% | 227.30 g/mol | C12H21NO3 | 5MG
SDP
Catalog No. 5000238935
Encompass_Preferred
N-octanoyl-L-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum-sensing signaling molecule used as an analytical standard and research reagent to study bacterial communication, gene regulation, and biofilm formation. It is supplied in solid form or as a 10 mM solution in DMSO; molecular formula C12H21NO3, molecular weight 227.30 g/mol, reported purity 99.40%.
  • Used as an analytical standard for quorum sensing studies.
  • High purity suitable for analytical and research applications.
  • Available as a solid and as a premade DMSO solution.
  • Well characterized molecular weight and formula for quantitative analysis.
  • Compatible with HPLC and mass spectrometry workflows.
Catalog No. 50-002-38935 Supplier Medchemexpress LLC Supplier No. HY124237AR5MG
N-octanoyl-L-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum-sensing signaling molecule used as an analytical standard and research reagent to study bacterial communication, gene regulation, and biofilm formation. It is supplied in solid form or as a 10 mM solution in DMSO; molecular formula C12H21NO3, molecular weight 227.30 g/mol, reported purity 99.40%.
  • Used as an analytical standard for quorum sensing studies.
  • High purity suitable for analytical and research applications.
  • Available as a solid and as a premade DMSO solution.
  • Well characterized molecular weight and formula for quantitative analysis.
  • Compatible with HPLC and mass spectrometry workflows.