Learn More
Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | MFCD0022730 | 99.4% | 227.30 | C12H21NO3 | 10MM 1ML
SDP
Catalog No. 5000239189
Encompass_Preferred
N-Octanoyl-L-homoserine lactone (C8-HSL) is a bacterial quorum-sensing signaling molecule used in microbiology and biochemical research to study cell-cell communication, modulation of virulence, and signaling pathways. Supplied as a 10 mM solution in DMSO, this compound is ready-to-use for in vitro assays and analytical methods.
  • High purity (99.4%)
  • Supplied as 10 mM solution in DMSO for convenient dosing
  • Suitable for quorum sensing and virulence studies
  • Compatible with HPLC, GC, and MS analytical methods
  • Stable small-molecule standard for assay development
  • Supplied in a 1 mL vial for small-volume experiments
Catalog No. 50-002-39189 Supplier Medchemexpress LLC Supplier No. HY124237A10MM 1ML
N-Octanoyl-L-homoserine lactone (C8-HSL) is a bacterial quorum-sensing signaling molecule used in microbiology and biochemical research to study cell-cell communication, modulation of virulence, and signaling pathways. Supplied as a 10 mM solution in DMSO, this compound is ready-to-use for in vitro assays and analytical methods.
  • High purity (99.4%)
  • Supplied as 10 mM solution in DMSO for convenient dosing
  • Suitable for quorum sensing and virulence studies
  • Compatible with HPLC, GC, and MS analytical methods
  • Stable small-molecule standard for assay development
  • Supplied in a 1 mL vial for small-volume experiments