Learn More
Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | MFCD11113139 | 99.4% | 227.30 | C12H21NO3 | 25MG
SDP
Catalog No. 5000236891
Encompass_Preferred
N-Octanoyl-L-homoserine lactone is a small, diffusible signaling molecule used in bacterial quorum sensing research. It is provided as a research-grade standard for studies of cell-cell communication, gene regulation, and virulence modulation. The product is available as a solid or as a DMSO solution and is intended for laboratory research use only.
Catalog No. 50-002-36891 Supplier Medchemexpress LLC Supplier No. HY124237A25MG
Add to Cart
Add to Cart
N-Octanoyl-L-homoserine lactone is a small, diffusible signaling molecule used in bacterial quorum sensing research. It is provided as a research-grade standard for studies of cell-cell communication, gene regulation, and virulence modulation. The product is available as a solid or as a DMSO solution and is intended for laboratory research use only.