Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | MFCD11113139 | 98.2% | 227.30 g·mol⁻1 | C12H21NO3 | 25MG
SDP

Catalog No. 5000238629
Encompass_Preferred

N-Octanoyl-L-homoserine lactone (C8-HSL) is a high-purity analytical standard used as a reference compound in quorum-sensing research and for analytical method development. Supplied as a stable solid, it is intended for assay calibration, method validation, and biochemical experiments that require a defined homoserine lactone standard.

  • High purity for reliable analytical results
  • Molecular formula C12H21NO3 and molecular weight 227.30 g·mol⁻1
  • Provided as a solid in a brown glass vial for improved stability
  • Suitable for assay calibration, method validation, and research applications
  • Store at -20°C and protect from light and moisture
  • Includes batch-specific certificate of analysis for traceability

Catalog No. 50-002-38629 Supplier Medchemexpress LLC Supplier No. HY124237AR25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Octanoyl-L-homoserine lactone (C8-HSL) is a high-purity analytical standard used as a reference compound in quorum-sensing research and for analytical method development. Supplied as a stable solid, it is intended for assay calibration, method validation, and biochemical experiments that require a defined homoserine lactone standard.

  • High purity for reliable analytical results
  • Molecular formula C12H21NO3 and molecular weight 227.30 g·mol⁻1
  • Provided as a solid in a brown glass vial for improved stability
  • Suitable for assay calibration, method validation, and research applications
  • Store at -20°C and protect from light and moisture
  • Includes batch-specific certificate of analysis for traceability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.