Learn More
Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | MFCD11113139 | 99.4% | 227.30 g/mol | C12H21NO3 | 10MG
SDP
Catalog No. 5000236889
Encompass_Preferred
N-Octanoyl-L-homoserine lactone is a small, diffusible acyl-homoserine lactone used as an analytical standard and research reagent for quorum sensing studies and bacterial communication assays. Supplied in high purity, it is suitable as an HPLC/LC-MS reference and for studies of virulence regulation and cell-cell signaling.
  • High purity (99.4%) for analytical applications
  • Available in solid and solution formats for flexible use
  • Molecular weight 227.30 g/mol and chemical formula C12H21NO3
  • Suitable for assay development and reference standards
Catalog No. 50-002-36889 Supplier Medchemexpress LLC Supplier No. HY124237A10MG
N-Octanoyl-L-homoserine lactone is a small, diffusible acyl-homoserine lactone used as an analytical standard and research reagent for quorum sensing studies and bacterial communication assays. Supplied in high purity, it is suitable as an HPLC/LC-MS reference and for studies of virulence regulation and cell-cell signaling.
  • High purity (99.4%) for analytical applications
  • Available in solid and solution formats for flexible use
  • Molecular weight 227.30 g/mol and chemical formula C12H21NO3
  • Suitable for assay development and reference standards