Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | MFCD11113139 | 99.4% | 227.30 g/mol | C12H21NO3 | 5MG
SDP

Catalog No. 5000238465
Encompass_Preferred

N-Octanoyl-L-homoserine lactone is a bacterial quorum-sensing signaling molecule used as an analytical standard and research reagent to study cell-cell communication, regulation of gene expression, and virulence. It is supplied as a high-purity solid suitable for biochemical assays, with small pack sizes and recommended DMSO solution preparations for experimental use.

  • High purity: 99.4% (reported)
  • CAS number: 147852-84-4
  • Molecular formula: C12H21NO3
  • Molecular weight: 227.30 g/mol
  • Available as small solid packs including 5 mg
  • Solution option: 10 mM in DMSO for immediate use

Catalog No. 50-002-38465 Supplier Medchemexpress LLC Supplier No. HY124237A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Octanoyl-L-homoserine lactone is a bacterial quorum-sensing signaling molecule used as an analytical standard and research reagent to study cell-cell communication, regulation of gene expression, and virulence. It is supplied as a high-purity solid suitable for biochemical assays, with small pack sizes and recommended DMSO solution preparations for experimental use.

  • High purity: 99.4% (reported)
  • CAS number: 147852-84-4
  • Molecular formula: C12H21NO3
  • Molecular weight: 227.30 g/mol
  • Available as small solid packs including 5 mg
  • Solution option: 10 mM in DMSO for immediate use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.