Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ n-Octanoylsucrose, Calbiochem™,

Catalog No. 4944665GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-446-65GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-446-65GM Supplier MilliporeSigma™ Supplier No. 4944665GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Quantity 5 g
Packaging Glass Bottle
Synonym n-Octanoyl-β-D-fructofuranosyl-α-D-glucopyranoside, Sucrose monocaproylate
Percent Purity ≥99% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.