Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 10mM 1mL | 29836-26-8 | 292.37 g/mol | C14H28O6 | 1 ML
SDP
Catalog No. 5000308047
Encompass_Preferred
n-Octyl β-D-glucopyranoside is a mild, non-ionic detergent used to solubilize membrane-bound proteins, prepare lipid vesicles, and aid membrane protein crystallization. It is supplied in solid and solution formats, including a ready-to-use 10 mM solution in DMSO (1 mL).
  • Solubilizes membrane proteins while preserving native structure.
  • Non-ionic, non-denaturing detergent suitable for protein reconstitution and crystallization.
  • Available as 10 mM solution in DMSO (1 mL) or various solid weights for scale-up.
  • Inhibits cavitation-induced cell lysis in vitro, aiding gentle sample handling.
Catalog No. 50-003-08047 Supplier Medchemexpress LLC Supplier No. HY11628510MM 1ML
Add to Cart
Add to Cart
n-Octyl β-D-glucopyranoside is a mild, non-ionic detergent used to solubilize membrane-bound proteins, prepare lipid vesicles, and aid membrane protein crystallization. It is supplied in solid and solution formats, including a ready-to-use 10 mM solution in DMSO (1 mL).
  • Solubilizes membrane proteins while preserving native structure.
  • Non-ionic, non-denaturing detergent suitable for protein reconstitution and crystallization.
  • Available as 10 mM solution in DMSO (1 mL) or various solid weights for scale-up.
  • Inhibits cavitation-induced cell lysis in vitro, aiding gentle sample handling.