Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 1mg | 869666-57-9 | 316.52 g·mol^-1 | C14H4D24O6 | 1 MG
SDP
Catalog No. 5000308046
Encompass_Preferred
n-Octyl β-D-glucopyranoside-d24 is a deuterium-labeled form of n-octyl β-D-glucopyranoside supplied as a 1 mg sample. It is intended for use as an isotopic tracer and internal standard in analytical workflows, with molecular formula C14H4D24O6 and molecular weight 316.52 g·mol⁻¹ (CAS 869666-57-9).
  • Deuterium-labeled (d24) for tracer and internal standard applications.
  • Designed for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High chemical and isotopic purity suitable for analytical workflows.
  • Nonionic glucoside structure useful for solubilizing membrane-associated molecules.
  • Supplied as a small, stable powder for convenient handling and storage.
Catalog No. 50-003-08046 Supplier Medchemexpress LLC Supplier No. HY116285S1MG
n-Octyl β-D-glucopyranoside-d24 is a deuterium-labeled form of n-octyl β-D-glucopyranoside supplied as a 1 mg sample. It is intended for use as an isotopic tracer and internal standard in analytical workflows, with molecular formula C14H4D24O6 and molecular weight 316.52 g·mol⁻¹ (CAS 869666-57-9).
  • Deuterium-labeled (d24) for tracer and internal standard applications.
  • Designed for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High chemical and isotopic purity suitable for analytical workflows.
  • Nonionic glucoside structure useful for solubilizing membrane-associated molecules.
  • Supplied as a small, stable powder for convenient handling and storage.