Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 5g | 29836-26-8 | 292.37 | C14H28O6 | 5 G
SDP
Catalog No. 5000229189
Encompass_Preferred
n-Octyl β-D-glucopyranoside is a non-ionic detergent used to solubilize and stabilize membrane proteins for biochemical and structural studies. It is commonly applied in membrane protein extraction, solubilization, reconstitution, and crystallization workflows, and can inhibit cavitation-induced cell lysis in vitro.
  • Mild, non-denaturing non-ionic detergent for membrane protein work.
  • Facilitates solubilization and reconstitution of membrane-bound proteins.
  • Suitable for membrane protein crystallization and structural analysis.
  • Can inhibit cavitation-induced cell lysis in vitro.
  • Powder is stable at refrigerated or frozen conditions for extended storage.
Catalog No. 50-002-29189 Supplier Medchemexpress LLC Supplier No. HY1162855G
n-Octyl β-D-glucopyranoside is a non-ionic detergent used to solubilize and stabilize membrane proteins for biochemical and structural studies. It is commonly applied in membrane protein extraction, solubilization, reconstitution, and crystallization workflows, and can inhibit cavitation-induced cell lysis in vitro.
  • Mild, non-denaturing non-ionic detergent for membrane protein work.
  • Facilitates solubilization and reconstitution of membrane-bound proteins.
  • Suitable for membrane protein crystallization and structural analysis.
  • Can inhibit cavitation-induced cell lysis in vitro.
  • Powder is stable at refrigerated or frozen conditions for extended storage.