Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 5mg | 869666-57-9 | 316.52 g/mol | C14H4D24O6 | 5 MG
SDP
Catalog No. 5000229854
Encompass_Preferred
n-Octyl β-D-glucopyranoside-d24 is a deuterium-labeled non-ionic glycoside reagent used as a stable isotope probe in biochemical and biophysical research. It is supplied as a purified solid for laboratory use (for research use only).
  • Deuterium-labeled (d24) for enhanced mass spectrometry and NMR analysis.
  • High purity (≈98.7%) suitable for analytical applications.
  • Available in small, research-scale quantities for method development.
  • Non-ionic glycoside structure useful as a membrane-mimetic detergent or probe.
  • Provided as a stable, characterized reagent with reported molecular weight and formula.
Catalog No. 50-002-29854 Supplier Medchemexpress LLC Supplier No. HY116285S5MG
n-Octyl β-D-glucopyranoside-d24 is a deuterium-labeled non-ionic glycoside reagent used as a stable isotope probe in biochemical and biophysical research. It is supplied as a purified solid for laboratory use (for research use only).
  • Deuterium-labeled (d24) for enhanced mass spectrometry and NMR analysis.
  • High purity (≈98.7%) suitable for analytical applications.
  • Available in small, research-scale quantities for method development.
  • Non-ionic glycoside structure useful as a membrane-mimetic detergent or probe.
  • Provided as a stable, characterized reagent with reported molecular weight and formula.