Learn More
Research Products International Corp n-Octylglucoside [n-Octyl-B-D-glucopyranoside], 10 Grams
SDP
Catalog No. 50213526
Encompass_Preferred
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Catalog No. 50-213-526 Supplier Research Products International Corp Supplier No. N0200710.0
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.