Learn More
Abcam n-Octyl-beta-D-glucopyranoside, Non-ionic detergent, 5G
Catalog No. 502583331
Encompass_Preferred
MW 292.37 Da. Non-ionic detergent. Solubilizes membrane-bound proteins. Permeabilizes cell membranes of paraformaldehyde fixed cells. Forms self-assembled nanostructures in water.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3331 Supplier Abcam Supplier No. AB1420715G
MW 292.37 Da. Non-ionic detergent. Solubilizes membrane-bound proteins. Permeabilizes cell membranes of paraformaldehyde fixed cells. Forms self-assembled nanostructures in water.
The product is subject to the following: Abcam Restricted Use Statement