Learn More
AdipoGen n-Octyl-beta-D-thioglucopyrano
SDP
Catalog No. 502562715
Encompass_Preferred
Chemical. CAS 85618-21-9. Formula C14H28O5S. MW 308.4. Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. The thioether linkage makes it resistant to degradation by beta-glucosidase enzymes. For the stabilization and activation of enzymes. Used for cell lysis experiments.
Catalog No. 50-256-2715 Supplier AdipoGen Supplier No. AGCC10005G005
Chemical. CAS 85618-21-9. Formula C14H28O5S. MW 308.4. Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. The thioether linkage makes it resistant to degradation by beta-glucosidase enzymes. For the stabilization and activation of enzymes. Used for cell lysis experiments.