Learn More
n-Octyl-β-D-glucopyranoside (White Crystalline Powder), Fisher BioReagents
Shop All Fisherbrand ProductsDescription
Specifications
Specifications
| Absorbance | 0.3 max. at 260nm |
| Color | White |
| Physical Form | Powder/Solid |
| Melting Point | 105°C |
| Quantity | 1 g |
| Packaging | Poly Micro Tube |
| Assay Percent Range | ≥98 % |
| Specific Rotation | -28.0 to -32.0° (+ or -) |
| Merck Index | 15, 6857 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Precautionary Statement | Emergency Overview The toxicological properties have not been fully investigated. NFPA Health: 1 Flammability: |
| Recommended Storage | -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.