Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl-trioxyethylene, Thermo Scientific™

Catalog No. AC124730050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC124730050 5 g
AC124730010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC124730050 Supplier Thermo Scientific Chemicals Supplier No. 124730050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19327-38-9
Molecular Formula C14H30O4
Molecular Weight (g/mol) 262.39
InChI Key XIVLVYLYOMHUGB-UHFFFAOYSA-N
Synonym triethylene glycol monooctyl ether, n-octyltrioxyethylene, octyltriglycol, n-octyl oxyethylene 3 ethanol, octyl triethylene glycol ether, 2-2-2-octoxyethoxy ethoxy ethanol, ooe 3e, 2-2-2-octyloxyethoxy ethoxy ethan-1-ol, n-octyl-trioxyethylene, triethyleneglycol octyl ether
PubChem CID 2723692
IUPAC Name 2-[2-(2-octoxyethoxy)ethoxy]ethanol
SMILES CCCCCCCCOCCOCCOCCO

Specifications

Density 0.94
Linear Formula CH3(CH2)7OCH2CH2OCH2CH2OCH2CH2OH
Quantity 5 g
Formula Weight 262.39
Chemical Name or Material n-Octyl-trioxyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.