Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. O04525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0452-5G 5 g
O04521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0452-5G Supplier TCI America Supplier No. O04525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28741-08-4
Molecular Formula C8H19BO2
Molecular Weight (g/mol) 158.048
MDL Number MFCD01074560
PubChem CID 5195177
IUPAC Name octylboronic acid
SMILES B(CCCCCCCC)(O)O

Specifications

Melting Point 81°C
Color White
Quantity 5 g
Synonym n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl
InChI Key octylboronic acid
Formula Weight 158.05
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.