Learn More
Research Products International Corp n-Octylglucoside [n-Octyl-B-D-glucopyranoside], 25 Grams
SDP
Supplier:  Research Products International Corp N0200725.0
Encompass_Preferred
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Catalog No. 50-488-838