Learn More
Sigma Aldrich Fine Chemicals Biosciences n Octylglucoside non ionic
Catalog No. 502903500
Encompass
1-O-n-Octyl--D-glucopyranoside octylglucoside is a detergent with an octyl chain as hydrophobic residue and glucose as hydrophilic part. It is a non-ionic dialyzable mild and non-denaturing detergent which is used for the solubilization isolation and reconstitution of membrane proteins.
Catalog No. 50-290-3500 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10634425001
1-O-n-Octyl--D-glucopyranoside octylglucoside is a detergent with an octyl chain as hydrophobic residue and glucose as hydrophilic part. It is a non-ionic dialyzable mild and non-denaturing detergent which is used for the solubilization isolation and reconstitution of membrane proteins.