Learn More
Medchemexpress LLC N-Octylmaleimide 10mM 1mL | 4080-76-6 | 209.28 | C12H19NO2 | 1 ML
SDP
Catalog No. 5000307660
Encompass_Preferred
N-Octylmaleimide (CAS 4080-76-6) is an alkylmaleimide reagent used in biochemical research. It is reported to inhibit rat liver glucose-6-phosphatase and is supplied in both solid forms and as a 10 mM solution in DMSO for laboratory use. The compound has molecular formula C12H19NO2 and molecular weight 209.28. For research use only.
  • Inhibits rat liver glucose-6-phosphatase.
  • Offered as solid and as 10 mM solution in DMSO.
  • Available in multiple solid mass sizes, including 10 mg-500 mg.
  • Molecular weight 209.28 and formula C12H19NO2.
  • Intended for research use only; not for patient use.
Catalog No. 50-003-07660 Supplier Medchemexpress LLC Supplier No. HY11575910MM 1ML
N-Octylmaleimide (CAS 4080-76-6) is an alkylmaleimide reagent used in biochemical research. It is reported to inhibit rat liver glucose-6-phosphatase and is supplied in both solid forms and as a 10 mM solution in DMSO for laboratory use. The compound has molecular formula C12H19NO2 and molecular weight 209.28. For research use only.
  • Inhibits rat liver glucose-6-phosphatase.
  • Offered as solid and as 10 mM solution in DMSO.
  • Available in multiple solid mass sizes, including 10 mg-500 mg.
  • Molecular weight 209.28 and formula C12H19NO2.
  • Intended for research use only; not for patient use.