Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyltriethoxysilane, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
2943-75-1
C14H32O3Si
276.49
MFCD00039883
MSRJTTSHWYDFIU-UHFFFAOYSA-N
n-octyltriethoxysilane,octyltriethoxysilane,triethoxy octyl silane,silane, triethoxyoctyl,dynasylan octeo,octyl triethoxy silane,prosil 9202,silquest a 137,prosil 9234,a 137 coupling agent
76262
triethoxy(octyl)silane
CCCCCCCC[Si](OCC)(OCC)OCC
2943-75-1
C14H32O3Si
276.49
MFCD00039883
MSRJTTSHWYDFIU-UHFFFAOYSA-N
n-octyltriethoxysilane,octyltriethoxysilane,triethoxy octyl silane,silane, triethoxyoctyl,dynasylan octeo,octyl triethoxy silane,prosil 9202,silquest a 137,prosil 9234,a 137 coupling agent
76262
triethoxy(octyl)silane
CCCCCCCC[Si](OCC)(OCC)OCC
Boiling Point 98°C
Flash Point 120°C
Specific Gravity 0.88
Refractive Index 1.4160 to 1.418
Color Colorless
Quantity 25mL
Infrared Spectrum Authentic
Density 0.8800g/mL
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)7Si(OCH2CH3)3
Solubility Information Solubility in water:
Formula Weight 276.49
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material n-Octyltriethoxysilane, 97%
Show More Show Less

GHS H Statement
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 220-941-2

RTECSNumber : VV6695500

TSCA : TSCA