Learn More
Cayman Chemical N-Oleoyl GlutamIn 1mg
SDP
Catalog No. 502939043
Encompass_Preferred
An endogenous N-acyl amine; induces mitochondrial uncoupling to increase respiration in C2C12 cells by 64% at 50 µM; inhibits capsaicin-induced calcium flux in HEK293A cells co-transfected with TRPV1 and RFP; reduces the number of acetic acid-induced abdominal constrictions, as well as formalin-induced paw licking in mice at 100 mg/kg
Catalog No. 50-293-9043 Supplier Cayman Chemical Supplier No. 289221 MG
An endogenous N-acyl amine; induces mitochondrial uncoupling to increase respiration in C2C12 cells by 64% at 50 µM; inhibits capsaicin-induced calcium flux in HEK293A cells co-transfected with TRPV1 and RFP; reduces the number of acetic acid-induced abdominal constrictions, as well as formalin-induced paw licking in mice at 100 mg/kg