Learn More
Cayman Chemical N-Oleoyl GlycIn 100mg
SDP
Supplier:  Cayman Chemical 90269100 MG
Encompass_Preferred
An endogenous lipoamino acid; increases triglyceride levels in 3T3-L1 adipocytes, an effect that can be reversed by SR141716; selectively induces the expression of mRNA encoding the CB1 receptor over mRNA encoding the CB2 receptor in the same cells at 50 µM; potentiates glycine-induced chloride currents in Xenopus oocytes expressing α1-, α1β-, α2-, α2β-, or α3 subunit-containing glycine receptors but not those expressing α3β subunit-containing glycine receptors at 1 µM; increases c-Fos protein levels, intracellular calcium levels, and AgRP secretion in mHypoE-38 cells, effects that can be reversed by AM251; reduces binge-like ethanol intake in alcohol-preferring mice without affecting total fluid intake and does not affect sucrose intake in the sucrose preference test in mice at 60 mg/kg; reduces balance impairments, thermal hyperalgesia, mechanical allodynia, aggressiveness, and immobility time in the tail suspension test in a mouse model of traumatic brain injury at 50 mg/kg
Catalog No. 50-289-2836