Learn More
Cayman Chemical N-Oleoyl-L-phenylalanine, Purity: ≥95%, CAS: 136560-78-6, 10 mg
SDP
Catalog No. 502940005
Encompass_Preferred
An N-acyl amide; increases the OCR of mitochondria isolated from mouse brown adipose tissue in a Seahorse assay and decreases oligomycin-induced increases in the mitochondrial membrane potential in C2C12 cells, indicating mitochondrial uncoupling activity at 50 µM; reduces food intake and body weight of diet-induced obese mice at 30 mg/kg for 8 days
Catalog No. 50-294-0005 Supplier Cayman Chemical Supplier No. 2892110 MG
An N-acyl amide; increases the OCR of mitochondria isolated from mouse brown adipose tissue in a Seahorse assay and decreases oligomycin-induced increases in the mitochondrial membrane potential in C2C12 cells, indicating mitochondrial uncoupling activity at 50 µM; reduces food intake and body weight of diet-induced obese mice at 30 mg/kg for 8 days