Learn More
Cayman Chemical N-Oleoyl-L-SerIn 1mg
SDP
Catalog No. 502955919
Encompass_Preferred
An endogenous lipid that has been reported to stimulate bone formation and to inhibit bone resorption
Catalog No. 50-295-5919 Supplier Cayman Chemical Supplier No. 130581 MG
An endogenous lipid that has been reported to stimulate bone formation and to inhibit bone resorption