Learn More
Cayman Chemical N-Oleoyl LeucIn 1mg
SDP
Catalog No. 502981517
Encompass_Preferred
An N-acyl amide that uncouples mitochondrial respiration independent of UCP1 in vitro; decreases body weight and food intake, preferentially decreasing fat mass in a diet-induced obesity mouse model; improves glucose homeostasis and increases energy expenditure while slightly decreasing overall locomotor activity
Catalog No. 50-298-1517 Supplier Cayman Chemical Supplier No. 200641 MG
An N-acyl amide that uncouples mitochondrial respiration independent of UCP1 in vitro; decreases body weight and food intake, preferentially decreasing fat mass in a diet-induced obesity mouse model; improves glucose homeostasis and increases energy expenditure while slightly decreasing overall locomotor activity