Learn More
Cayman Chemical N-Oleoyl ProlIn 10mg
SDP
Catalog No. 502981522
Encompass_Preferred
An endogenous N-acyl amine that is decreased in lumbar spinal cord after capsaicin administration (1 UG/10 µl) and has surfactant properties (critical micelle concentration = 4.8 µM)
Catalog No. 50-298-1522 Supplier Cayman Chemical Supplier No. 2036910 MG
An endogenous N-acyl amine that is decreased in lumbar spinal cord after capsaicin administration (1 UG/10 µl) and has surfactant properties (critical micelle concentration = 4.8 µM)