Learn More
Cayman Chemical N-Oleoyl Proline, Purity: ≥95%, CAS: 107432-37-1, 1 mg
SDP
Catalog No. 502981772
Encompass_Preferred
An endogenous N-acyl amine that is decreased in lumbar spinal cord after capsaicin administration (1 µg/10 µl) and has surfactant properties (critical micelle concentration = 4.8 µM)
Catalog No. 50-298-1772 Supplier Cayman Chemical Supplier No. 203691 MG
An endogenous N-acyl amine that is decreased in lumbar spinal cord after capsaicin administration (1 µg/10 µl) and has surfactant properties (critical micelle concentration = 4.8 µM)