Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-oleoyl taurine | 52514-04-2 | 99.9% | 1 MG
SDP

Catalog No. 5000233975
Encompass_Preferred

N-oleoyl taurine is a fatty acid-taurine conjugate supplied as a DMSO solution for research use. It is described as an endogenous metabolite with reported activity at TRPV1 and TRPV4, and is provided with certificate of analysis (COA) and selectable batches.

  • Supplied as a solution in DMSO (1 mg: 12.83 mM · 200 μL)
  • High purity (reported 99.9%)
  • Molecular weight 389.59 g/mol and formula C20H39NO4S
  • Stored as a solution at -20°C for up to 2 years
  • COA and batch information available for traceability

Catalog No. 50-002-33975 Supplier Medchemexpress LLC Supplier No. HY1209631MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-oleoyl taurine is a fatty acid-taurine conjugate supplied as a DMSO solution for research use. It is described as an endogenous metabolite with reported activity at TRPV1 and TRPV4, and is provided with certificate of analysis (COA) and selectable batches.

  • Supplied as a solution in DMSO (1 mg: 12.83 mM · 200 μL)
  • High purity (reported 99.9%)
  • Molecular weight 389.59 g/mol and formula C20H39NO4S
  • Stored as a solution at -20°C for up to 2 years
  • COA and batch information available for traceability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.